missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(hydroxymethyl)benzeneboronic acid, 98%

CAS: 481681-02-1 | C7H8BFO3 | 169.95 g/mol

Supplier:  Thermo Scientific Chemicals H5248703

 View more versions of this product

Catalog No. AAH5248703

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluoro-3-(hydroxymethyl)benzeneboronic acid
124°C to 126°C
MFCD08235075
4-fluoro-3-hydroxymethyl phenyl boronic acid, 4-fluoro-3-hydroxymethyl phenylboronic acid, 4-fluoro-3-hydroxymethyl benzeneboronic acid, 4-fluoro-3-hydroxymethylphenylboronic acid, boronic acid, 4-fluoro-3-hydroxymethyl phenyl, boronic acid,b-4-fluoro-3-hydroxymethyl phenyl, pubchem23767, acmc-209kcj, ksc496e1t
OCC1=C(F)C=CC(=C1)B(O)O
169.95
169.95
481681-02-1
C7H8BFO3
1 g
PWMOQHMTXJYUGE-UHFFFAOYSA-N
[4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
21865588
98%

Chemical Identifiers

481681-02-1
169.95
PWMOQHMTXJYUGE-UHFFFAOYSA-N
21865588
OCC1=C(F)C=CC(=C1)B(O)O
C7H8BFO3
MFCD08235075
4-fluoro-3-hydroxymethyl phenyl boronic acid, 4-fluoro-3-hydroxymethyl phenylboronic acid, 4-fluoro-3-hydroxymethyl benzeneboronic acid, 4-fluoro-3-hydroxymethylphenylboronic acid, boronic acid, 4-fluoro-3-hydroxymethyl phenyl, boronic acid,b-4-fluoro-3-hydroxymethyl phenyl, pubchem23767, acmc-209kcj, ksc496e1t
[4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only