missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(hydroxymethyl)benzeneboronic acid, 98%

Catalog No. AAH52487MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52487MD 250 mg
AAH5248703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52487MD Supplier Thermo Scientific Chemicals Supplier No. H52487MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 481681-02-1 | C7H8BFO3 | 169.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 481681-02-1
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD08235075
InChI Key PWMOQHMTXJYUGE-UHFFFAOYSA-N
Synonym 4-fluoro-3-hydroxymethyl phenyl boronic acid, 4-fluoro-3-hydroxymethyl phenylboronic acid, 4-fluoro-3-hydroxymethyl benzeneboronic acid, 4-fluoro-3-hydroxymethylphenylboronic acid, boronic acid, 4-fluoro-3-hydroxymethyl phenyl, boronic acid,b-4-fluoro-3-hydroxymethyl phenyl, pubchem23767, acmc-209kcj, ksc496e1t
PubChem CID 21865588
IUPAC Name [4-fluoro-3-(hydroxymethyl)phenyl]boronic acid
SMILES OCC1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 124°C to 126°C
Quantity 250 mg
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-(hydroxymethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only