missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. p-7129607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03091G 1 g
F03095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03091G Supplier TCI America Supplier No. F03091G
Only null left

Chemical Identifiers

CAS 194853-86-6
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.11
MDL Number MFCD00061283
InChI Key LCCPQUYXMFXCAC-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-fluorobenzonitrile, 4-fluoro-2-trifluoromethyl-benzonitrile, 2-cyano-5-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolunitrile, benzonitrile, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzenecarbonitrile, pubchem4802
PubChem CID 605085
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzonitrile
SMILES FC1=CC=C(C#N)C(=C1)C(F)(F)F

Specifications

Melting Point 46°C
Quantity 1 g
Formula Weight 189.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzonitrile