missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzonitrile, 98+%

Catalog No. AAB2061714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2061714 25 g
AAB2061706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2061714 Supplier Thermo Scientific Chemicals Supplier No. B2061714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 194853-86-6 | C8H3F4N | 189.11 g/mol

It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 194853-86-6
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.11
MDL Number MFCD00061283
InChI Key LCCPQUYXMFXCAC-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzonitrile, 2-trifluoromethyl-4-fluorobenzonitrile, 4-fluoro-2-trifluoromethyl-benzonitrile, 2-cyano-5-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolunitrile, benzonitrile, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzenecarbonitrile, pubchem4802
PubChem CID 605085
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzonitrile
SMILES FC1=CC=C(C#N)C(=C1)C(F)(F)F

Specifications

Melting Point 43°C to 45°C
Density 1.35
Boiling Point 194°C to 196°C
Quantity 25 g
UN Number UN3439
Solubility Information Insoluble in water. Solubility in methanol is almost transparent.
Formula Weight 189.11
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only