missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzoic Acid 97.0+%, TCI America™

Catalog No. F05771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05771G 1 g
F05775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05771G Supplier TCI America Supplier No. F05771G
Only null left

Chemical Identifiers

CAS 141179-72-8
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040982
InChI Key JUHPDXOIGLHXTC-UHFFFAOYSA-M
Synonym 4-fluoro-2-trifluoromethyl benzoic acid, 4-fluoro-2 trifluoromethyl benzoic acid, alpha,alpha,alpha,4-tetrafluoro-o-toluic acid, benzoic acid, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl benzoicacid, pubchem4971, acmc-209clt, intermediates-zcf02077, ksc183o7t
PubChem CID 688255
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzoate
SMILES [O-]C(=O)C1=CC=C(F)C=C1C(F)(F)F

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoic Acid