missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzaldehyde 98.0+%, TCI America™

Catalog No. F05441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05441G 1 g
F05445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05441G Supplier TCI America Supplier No. F05441G
Only null left

Chemical Identifiers

CAS 90176-80-0
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
InChI Key NONOHEMDNFTKCZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzaldehyde, 2-trifluoromethyl-4-fluorobenzaldehyde, 4-fluoro-2-trifluoromethyl-benzaldehyde, benzaldehyde, 4-fluoro-2-trifluoromethyl, 5-fluoro-2-formylbenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-o-tolualdehyde, pubchem4241, ksc487q2t, paragos 530044
PubChem CID 522940
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1F)C(F)(F)F)C=O

Specifications

Color Yellow
Boiling Point 114°C
Quantity 1 g
UN Number 1989
Formula Weight 192.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzaldehyde