missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-nitrotoluene 96.0+%, TCI America™
Supplier: TCI America F02185G
Specifications
| 4-Fluoro-2-nitrotoluene | |
| 139°C | |
| MFCD00007200 | |
| 4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482 | |
| CC1=C(C=C(C=C1)F)[N+](=O)[O-] | |
| 155.128 | |
| 155.13 | |
| Liquid |
| 446-10-6 | |
| C7H6FNO2 | |
| 5 g | |
| SKWTUNAAJNDEIK-UHFFFAOYSA-N | |
| 4-fluoro-1-methyl-2-nitrobenzene | |
| 67965 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 446-10-6 | |
| 155.128 | |
| SKWTUNAAJNDEIK-UHFFFAOYSA-N | |
| 67965 | |
| CC1=C(C=C(C=C1)F)[N+](=O)[O-] |
| C7H6FNO2 | |
| MFCD00007200 | |
| 4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482 | |
| 4-fluoro-1-methyl-2-nitrobenzene |
Safety and Handling
TSCA : No