missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrotoluene 96.0+%, TCI America™

Supplier:  TCI America F021825G

 View more versions of this product

Catalog No. F021825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Fluoro-2-nitrotoluene
139°C
MFCD00007200
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
155.128
155.13
Liquid
446-10-6
C7H6FNO2
25 g
SKWTUNAAJNDEIK-UHFFFAOYSA-N
4-fluoro-1-methyl-2-nitrobenzene
67965
≥96.0% (GC)

Chemical Identifiers

446-10-6
155.128
SKWTUNAAJNDEIK-UHFFFAOYSA-N
67965
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
C7H6FNO2
MFCD00007200
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
4-fluoro-1-methyl-2-nitrobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions