missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$171.34 - $171.34
Specifications
| CAS | 139911-29-8 |
|---|---|
| Melting Point | 201°C |
| Color | White-Yellow |
| Molecular Formula | C7H8BFO2 |
| MDL Number | MFCD02093072 |
Specifications
| 139911-29-8 | |
| White-Yellow | |
| MFCD02093072 | |
| B(C1=C(C=C(C=C1)F)C)(O)O | |
| 153.947 | |
| 153.95 |
| 201°C | |
| C7H8BFO2 | |
| IQMLIVUHMSIOQP-UHFFFAOYSA-N | |
| (4-fluoro-2-methylphenyl)boronic acid | |
| 2734665 | |
| Crystalline Powder |