missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F06661G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06661G Supplier TCI America Supplier No. F06661G

Specifications

Chemical Name or Material 4-Fluoro-2-methylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 139911-29-8
Melting Point 201°C
Color White-Yellow
Molecular Formula C7H8BFO2
Quantity 1 g
MDL Number MFCD02093072
Synonym 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp
InChI Key IQMLIVUHMSIOQP-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)F)C)(O)O
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 153.947
PubChem CID 2734665
Formula Weight 153.95
Physical Form Crystalline Powder
Show More Show Less