missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzyl alcohol, 99%

Catalog No. p-7031328
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3236403 1 g
AAH3236406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3236403 Supplier Thermo Scientific Chemicals Supplier No. H3236403
Only null left

CAS: 80141-91-9 | C8H9FO | 140.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80141-91-9
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD03701058
InChI Key YSULUXOLTMBSFF-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylphenyl methanol, 4-fluoro-2-methylbenzyl alcohol, 4-fluoro-2-methylbenzylalcohol, rarechem al bd 0506, 4-fluoro-2-methylbenzenemethanol, benzenemethanol, 4-fluoro-2-methyl, 4-fluoro-2-methyl-phenyl-methanol, 4-fluoro-2-methylphenyl methan-1-ol, 4'-fluoro-2-methylbenzyl alcohol
PubChem CID 3872017
IUPAC Name (4-fluoro-2-methylphenyl)methanol
SMILES CC1=C(C=CC(=C1)F)CO

Specifications

Quantity 1 g
Formula Weight 140.16
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methylbenzyl alcohol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only