missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methylbenzonitrile 99.0+%, TCI America™
$54.06 - $214.37
Chemical Identifiers
| CAS | 147754-12-9 |
|---|---|
| Molecular Formula | C8H6FN |
| Molecular Weight (g/mol) | 135.141 |
| MDL Number | MFCD03095106 |
| InChI Key | BJBXUIUJKPOZLV-UHFFFAOYSA-N |
| Synonym | 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l |
| PubChem CID | 3826055 |
| IUPAC Name | 4-fluoro-2-methylbenzonitrile |
| SMILES | CC1=C(C=CC(=C1)F)C#N |
Chemical Identifiers
| 147754-12-9 | |
| 135.141 | |
| BJBXUIUJKPOZLV-UHFFFAOYSA-N | |
| 3826055 | |
| CC1=C(C=CC(=C1)F)C#N |
| C8H6FN | |
| MFCD03095106 | |
| 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l | |
| 4-fluoro-2-methylbenzonitrile |
Specifications
| 147754-12-9 | |
| White | |
| MFCD03095106 | |
| 3439 | |
| BJBXUIUJKPOZLV-UHFFFAOYSA-N | |
| 4-fluoro-2-methylbenzonitrile | |
| 3826055 | |
| ≥99.0% (GC) | |
| 4-Fluoro-2-methylbenzonitrile |
| 71°C | |
| C8H6FN | |
| 1 g | |
| 2-methyl-4-fluorobenzonitrile, 4-fluoro-2-methyl-benzonitrile, benzonitrile, 4-fluoro-2-methyl, 2-cyano-5-fluorotoluene, 4-fluoro-2-methylbenzenecarbonitrile, pubchem1554, intermediates-zcf02129, methyl 4-fluorobenzonitrile, 4-fuoro-2-methylbenzonitrile, ksc174s6l | |
| CC1=C(C=CC(=C1)F)C#N | |
| 135.141 | |
| 135.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No