Learn More
4-Fluoro-2-methylbenzeneboronic acid, 98%
CAS: 139911-29-8 | C7H8BFO2 | 153.947 g/mol
Supplier: Thermo Scientific Chemicals B2411706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Fluoro-2-methylbenzeneboronic acid | |
| 199°C to 201°C | |
| MFCD02093072 | |
| 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp | |
| B(C1=C(C=C(C=C1)F)C)(O)O | |
| 153.947 | |
| 153.95 |
| 139911-29-8 | |
| C7H8BFO2 | |
| 5 g | |
| IQMLIVUHMSIOQP-UHFFFAOYSA-N | |
| (4-fluoro-2-methylphenyl)boronic acid | |
| 2734665 | |
| 98% |
Chemical Identifiers
| 139911-29-8 | |
| 153.947 | |
| IQMLIVUHMSIOQP-UHFFFAOYSA-N | |
| 2734665 | |
| B(C1=C(C=C(C=C1)F)C)(O)O |
| C7H8BFO2 | |
| MFCD02093072 | |
| 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp | |
| (4-fluoro-2-methylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only