missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzeneboronic acid, 98%

Catalog No. AAB2411703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2411703 1 g
AAB2411706 5 g
AAB2411714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2411703 Supplier Thermo Scientific Chemicals Supplier No. B2411703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 139911-29-8 | C7H8BFO2 | 153.947 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139911-29-8
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.947
MDL Number MFCD02093072
InChI Key IQMLIVUHMSIOQP-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp
PubChem CID 2734665
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)F)C)(O)O

Specifications

Melting Point 199°C to 201°C
Quantity 1 g
Formula Weight 153.95
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only