missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methylbenzaldehyde 95.0+%, TCI America™
$37.29 - $82.02
Chemical Identifiers
| CAS | 63082-45-1 |
|---|---|
| Molecular Formula | C8H7FO |
| Molecular Weight (g/mol) | 138.141 |
| MDL Number | MFCD02261764 |
| InChI Key | ADCFIKGEGWFWEA-UHFFFAOYSA-N |
| Synonym | 4-fluoro-o-tolualdehyde, 4-fluoro-2-methyl benzaldehyde, 2-methyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methyl, 4-fluoro-2-methyl-benzaldehyde, pubchem1456, 5-fluoro-2-formyltoluene, acmc-1b7wn, ksc352s3j, 4-fluoro-2-methylbenzaldehyde? |
| PubChem CID | 2783217 |
| IUPAC Name | 4-fluoro-2-methylbenzaldehyde |
| SMILES | CC1=C(C=CC(=C1)F)C=O |
Chemical Identifiers
| 63082-45-1 | |
| 138.141 | |
| ADCFIKGEGWFWEA-UHFFFAOYSA-N | |
| 2783217 | |
| CC1=C(C=CC(=C1)F)C=O |
| C8H7FO | |
| MFCD02261764 | |
| 4-fluoro-o-tolualdehyde, 4-fluoro-2-methyl benzaldehyde, 2-methyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methyl, 4-fluoro-2-methyl-benzaldehyde, pubchem1456, 5-fluoro-2-formyltoluene, acmc-1b7wn, ksc352s3j, 4-fluoro-2-methylbenzaldehyde? | |
| 4-fluoro-2-methylbenzaldehyde |
Specifications
| 63082-45-1 | |
| 55°C | |
| MFCD02261764 | |
| 4-fluoro-o-tolualdehyde, 4-fluoro-2-methyl benzaldehyde, 2-methyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methyl, 4-fluoro-2-methyl-benzaldehyde, pubchem1456, 5-fluoro-2-formyltoluene, acmc-1b7wn, ksc352s3j, 4-fluoro-2-methylbenzaldehyde? | |
| CC1=C(C=CC(=C1)F)C=O | |
| 138.141 | |
| 138.14 | |
| Liquid |
| Yellow | |
| C8H7FO | |
| 1 g | |
| ADCFIKGEGWFWEA-UHFFFAOYSA-N | |
| 4-fluoro-2-methylbenzaldehyde | |
| 2783217 | |
| ≥95.0% (GC) | |
| 4-Fluoro-2-methylbenzaldehyde |
Safety and Handling
TSCA : No