missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzylamine, 97%

CAS: 870563-60-3 | C8H10FNO | 155.17 g/mol

$248.90 - $810.27

Chemical Identifiers

CAS 870563-60-3
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.17
MDL Number MFCD04116361
InChI Key OUYQGZMPYJPPER-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxybenzylamine, 2-methoxy-4-fluorobenzylamine, benzenemethanamine, 4-fluoro-2-methoxy, 1-4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxy-benzylamine
PubChem CID 17750694
IUPAC Name (4-fluoro-2-methoxyphenyl)methanamine
SMILES COC1=C(CN)C=CC(F)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2679603
View Documents
Thermo Scientific Chemicals
H2679603
1 g
Each for $248.90
Only null left
 
AAH2679606
View Documents
Thermo Scientific Chemicals
H2679606
5 g
Each for $810.27
Only null left
 
Description

Description

As pharmaceutical intermediate. As a metabolite of Capsaicin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As pharmaceutical intermediate. As a metabolite of Capsaicin.

Solubility
Insoluble in water.

Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications

Chemical Identifiers

870563-60-3
155.17
OUYQGZMPYJPPER-UHFFFAOYSA-N
17750694
COC1=C(CN)C=CC(F)=C1
C8H10FNO
MFCD04116361
4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxybenzylamine, 2-methoxy-4-fluorobenzylamine, benzenemethanamine, 4-fluoro-2-methoxy, 1-4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxy-benzylamine
(4-fluoro-2-methoxyphenyl)methanamine

Specifications

870563-60-3
1.524
1 g
Air Sensitive
Insoluble in water.
COC1=C(CN)C=CC(F)=C1
155.17
155.17
4-Fluoro-2-methoxybenzylamine
C8H10FNO
MFCD04116361
UN2735
4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxybenzylamine, 2-methoxy-4-fluorobenzylamine, benzenemethanamine, 4-fluoro-2-methoxy, 1-4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxy-benzylamine
OUYQGZMPYJPPER-UHFFFAOYSA-N
(4-fluoro-2-methoxyphenyl)methanamine
17750694
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only