missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzylamine, 97%

Catalog No. AAH2679603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2679603 1 g
AAH2679606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2679603 Supplier Thermo Scientific Chemicals Supplier No. H2679603

CAS: 870563-60-3 | C8H10FNO | 155.17 g/mol

As pharmaceutical intermediate. As a metabolite of Capsaicin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As pharmaceutical intermediate. As a metabolite of Capsaicin.

Solubility
Insoluble in water.

Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 870563-60-3
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.17
MDL Number MFCD04116361
InChI Key OUYQGZMPYJPPER-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxybenzylamine, 2-methoxy-4-fluorobenzylamine, benzenemethanamine, 4-fluoro-2-methoxy, 1-4-fluoro-2-methoxyphenyl methanamine, 4-fluoro-2-methoxy-benzylamine
PubChem CID 17750694
IUPAC Name (4-fluoro-2-methoxyphenyl)methanamine
SMILES COC1=C(CN)C=CC(F)=C1

Specifications

Refractive Index 1.524
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 155.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxybenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only