missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzonitrile, 97%

Catalog No. AAH2662703
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2662703 1 g
AAH2662709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2662703 Supplier Thermo Scientific Chemicals Supplier No. H2662703

CAS: 191014-55-8 | C8H6FNO | 151.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 191014-55-8
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD04116335
InChI Key HGBKZVIQHCUHRI-UHFFFAOYSA-N
Synonym benzonitrile, 4-fluoro-2-methoxy, 2-methoxy-4-fluorobenzonitrile, 4-fluoro-2-methoxy-benzonitrile, benzonitrile, 4-fluoro-2-methoxy-9ci, 4-fluoro-2-methoxybenzenecarbonitrile, pubchem4786, acmc-209xzr, ksc494i6t, 4fluoro-2-methoxy-benzonitrile, buttpark 80\07-22
PubChem CID 2783329
IUPAC Name 4-fluoro-2-methoxybenzonitrile
SMILES COC1=C(C=CC(=C1)F)C#N

Specifications

Melting Point 75°C to 77°C
Quantity 1 g
UN Number UN3439
Formula Weight 151.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only