missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. F06801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06801G 1 g
F06805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06801G Supplier TCI America Supplier No. F06801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 395-82-4
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00673010
InChI Key UUQDNAPKWPKHMK-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluorobenzoic acid, 4-fluoro-2-methoxy-benzoic acid, 4-fluoro-o-anisic acid, benzoic acid, 4-fluoro-2-methoxy, pubchem4969, acmc-209j6b, ksc494s1h, 4-fluoro-2-methoxybenzoicacid, rarechem al be 1023, 4-fluoro-2-methoxy benzoic acid
PubChem CID 2774545
IUPAC Name 4-fluoro-2-methoxybenzoic acid
SMILES COC1=CC(F)=CC=C1C(O)=O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 1 g
Formula Weight 170.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-methoxybenzoic Acid