missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxybenzoic acid, 99%

Catalog No. AAH2617906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2617906 5 g
AAH2617903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2617906 Supplier Thermo Scientific Chemicals Supplier No. H2617906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 395-82-4 | C8H7FO3 | 170.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 395-82-4
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00673010
InChI Key UUQDNAPKWPKHMK-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluorobenzoic acid, 4-fluoro-2-methoxy-benzoic acid, 4-fluoro-o-anisic acid, benzoic acid, 4-fluoro-2-methoxy, pubchem4969, acmc-209j6b, ksc494s1h, 4-fluoro-2-methoxybenzoicacid, rarechem al be 1023, 4-fluoro-2-methoxy benzoic acid
PubChem CID 2774545
IUPAC Name 4-fluoro-2-methoxybenzoic acid
SMILES COC1=CC(F)=CC=C1C(O)=O

Specifications

Melting Point 134°C to 136°C
Quantity 5 g
Formula Weight 170.14
Percent Purity 99%
Chemical Name or Material 4-Fluoro-2-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only