missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxyaniline 98.0+%, TCI America™

Catalog No. F10031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10031G 1 g
F10035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10031G Supplier TCI America Supplier No. F10031G

Chemical Identifiers

CAS 450-91-9
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.15
MDL Number MFCD00077536
InChI Key BNRRMRUVYDETQC-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluoroaniline, 2-amino-5-fluoroanisole, 4-fluoro-2-methoxybenzenamine, benzenamine, 4-fluoro-2-methoxy, 2-amino-5-fluoroanisole 4-fluoro-2-methoxyaniline, 4-fluoro-o-anisidine, acmc-1aegy, 4-fluoro-2-methoxy-aniline, ksc798g5t, 4-fluoro-2-methoxyphenylamine
PubChem CID 13532228
IUPAC Name 4-fluoro-2-methoxyaniline
SMILES COC1=C(N)C=CC(F)=C1

Specifications

Boiling Point 215°C
Quantity 1 g
UN Number 2810
Formula Weight 141.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-methoxyaniline