missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methoxyaniline, 95%

Catalog No. AAH6196503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6196503 1 g
AAH6196506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6196503 Supplier Thermo Scientific Chemicals Supplier No. H6196503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 450-91-9 | C7H8FNO | 141.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 450-91-9
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.15
MDL Number MFCD00077536
InChI Key BNRRMRUVYDETQC-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluoroaniline, 2-amino-5-fluoroanisole, 4-fluoro-2-methoxybenzenamine, benzenamine, 4-fluoro-2-methoxy, 2-amino-5-fluoroanisole 4-fluoro-2-methoxyaniline, 4-fluoro-o-anisidine, acmc-1aegy, 4-fluoro-2-methoxy-aniline, ksc798g5t, 4-fluoro-2-methoxyphenylamine
PubChem CID 13532228
IUPAC Name 4-fluoro-2-methoxyaniline
SMILES COC1=C(N)C=CC(F)=C1

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 141.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-methoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only