missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-iodotoluene 97.0+%, TCI America™

Catalog No. F06135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06135G 5 g
F061325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06135G Supplier TCI America Supplier No. F06135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13194-67-7
Molecular Formula C7H6FI
Molecular Weight (g/mol) 236.028
MDL Number MFCD00013709
InChI Key RZGYAMQMAVTAKP-UHFFFAOYSA-N
Synonym 4-fluoro-2-iodotoluene, benzene, 4-fluoro-2-iodo-1-methyl, 2-iodo-4-fluorotoluene, 4-fluoro-2-iodo-1-methyl-benzene, 1-fluoro-3-iodo-4-methyl-benzene, 4-fluor-2-iod-1-methylbenzol, zlchem 432, pubchem1622, acmc-209bo9
PubChem CID 83220
IUPAC Name 4-fluoro-2-iodo-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)I

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 236.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-iodotoluene