missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,6-dinitrophenol, 98%

Catalog No. AAA1730903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1730903 1 g
AAA1730906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1730903 Supplier Thermo Scientific Chemicals Supplier No. A1730903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 364-32-9 | C6H3FN2O5 | 202.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 364-32-9
Molecular Formula C6H3FN2O5
Molecular Weight (g/mol) 202.10
MDL Number MFCD00179333
InChI Key MDOWEUXXLVBZIU-UHFFFAOYSA-N
Synonym 4-fluoro-2,6-dinitrophenol, 2,6-dinitro-4-fluorophenol, phenol, 4-fluoro-2,6-dinitro, acmc-20ao7v, phenol, 2,6-dinitro-4-fluoro-,
PubChem CID 94951
IUPAC Name 4-fluoro-2,6-dinitrophenol
SMILES OC1=C(C=C(F)C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 50°C to 56°C
Quantity 1 g
Formula Weight 202.1
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2,6-dinitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only