Learn More
4-Fluoro-2,3-dimethylaniline, 97%
CAS: 1737-68-4 | C8H10FN | 139.173 g/mol
Supplier: Thermo Scientific Chemicals H3156003
Description
4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
| 4-Fluoro-2,3-dimethylaniline | |
| 1737-68-4 | |
| C8H10FN | |
| UN2811 | |
| LKMJLYCMLAPNDB-UHFFFAOYSA-N | |
| 4-fluoro-2,3-dimethylaniline | |
| 17770833 | |
| 97% |
| 42°C to 46°C | |
| 1 g | |
| MFCD04972870 | |
| 3-amino-6-fluoro-1,2-dimethylbenzene, 4-fluoro-2,3-dimethyl aniline, 2,3-dimethyl-4-fluoroaniline, 4-fluoro-o-xylidine, benzenamine, 4-fluoro-2,3-dimethyl, 4-fluoro-2,3-dimethyl-phenylamine, p-fluordimethylanilin, intermediates-zcf02293, 4-fluoranyl-2,3-dimethyl-aniline, 4-fluoro-2,3-dimethyl-benzenamine | |
| CC1=C(C=CC(=C1C)F)N | |
| 139.173 | |
| 139.17 |
Chemical Identifiers
| 1737-68-4 | |
| 139.173 | |
| LKMJLYCMLAPNDB-UHFFFAOYSA-N | |
| 17770833 | |
| CC1=C(C=CC(=C1C)F)N |
| C8H10FN | |
| MFCD04972870 | |
| 3-amino-6-fluoro-1,2-dimethylbenzene, 4-fluoro-2,3-dimethyl aniline, 2,3-dimethyl-4-fluoroaniline, 4-fluoro-o-xylidine, benzenamine, 4-fluoro-2,3-dimethyl, 4-fluoro-2,3-dimethyl-phenylamine, p-fluordimethylanilin, intermediates-zcf02293, 4-fluoranyl-2,3-dimethyl-aniline, 4-fluoro-2,3-dimethyl-benzenamine | |
| 4-fluoro-2,3-dimethylaniline |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only