missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,3-dimethylaniline, 97%

Catalog No. AAH31560MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31560MD 250 mg
AAH3156003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31560MD Supplier Thermo Scientific Chemicals Supplier No. H31560MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1737-68-4 | C8H10FN | 139.173 g/mol

4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1737-68-4
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD04972870
InChI Key LKMJLYCMLAPNDB-UHFFFAOYSA-N
Synonym 3-amino-6-fluoro-1,2-dimethylbenzene, 4-fluoro-2,3-dimethyl aniline, 2,3-dimethyl-4-fluoroaniline, 4-fluoro-o-xylidine, benzenamine, 4-fluoro-2,3-dimethyl, 4-fluoro-2,3-dimethyl-phenylamine, p-fluordimethylanilin, intermediates-zcf02293, 4-fluoranyl-2,3-dimethyl-aniline, 4-fluoro-2,3-dimethyl-benzenamine
PubChem CID 17770833
IUPAC Name 4-fluoro-2,3-dimethylaniline
SMILES CC1=C(C=CC(=C1C)F)N

Specifications

Melting Point 42°C to 46°C
Quantity 250 mg
UN Number UN2811
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2,3-dimethylaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only