missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1H-imidazole 97.0+%, TCI America™

Catalog No. F11571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F11571G Supplier TCI America Supplier No. F11571G
Only null left

Chemical Identifiers

CAS 30086-17-0
Molecular Formula C3H3FN2
Molecular Weight (g/mol) 86.07
MDL Number MFCD09835574
InChI Key HWUPTWDARIVICI-UHFFFAOYSA-N
PubChem CID 99297
IUPAC Name 5-fluoro-1H-imidazole
SMILES FC1=CN=CN1

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 86.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-1H-imidazole
TSCA No
Recommended Storage Refrigerator