accessibility menu, dialog, popup

UserName

CAS RN 30086-17-0

FNHN

CAS RN 30086-17-0

IUPAC Name: 5-fluoro-1H-imidazole
Synonyms: 4-fluoro-3H-imidazole
Molecular Weight (g/mol): 86.07
Molecular Formula: C3H3FN2
InChi Key: HWUPTWDARIVICI-UHFFFAOYSA-N
SMILES: FC1=CN=CN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results