missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-naphthoic acid, 97%

CAS: 573-03-5 | C11H7FO2 | 190.173 g/mol

$93.04 - $310.70

Chemical Identifiers

CAS 573-03-5
Molecular Formula C11H7FO2
Molecular Weight (g/mol) 190.173
MDL Number MFCD00004011
InChI Key DEWIOKQDRWFLFW-UHFFFAOYSA-N
Synonym 4-fluoro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 4-fluoro, 4-fluoronaphthalenecarboxylic acid, 4-fluoro-1-naphthalenecarboxylic acid, 4-fluoro-1-naphthalene carboxylic acid, 4-fluoro-naphthalene-1-carboxylic acid, acmc-1atch, maybridge3_003028, ksc270s4j, 1-carboxy-4-fluoronaphthalene
PubChem CID 68448
IUPAC Name 4-fluoronaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=C2F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1677803
View Documents
Thermo Scientific Chemicals
A1677803
1 g
Each for $93.04
Only null left
 
AAA1677806
View Documents
Thermo Scientific Chemicals
A1677806
5 g
Each for $310.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

573-03-5
190.173
DEWIOKQDRWFLFW-UHFFFAOYSA-N
68448
C1=CC=C2C(=C1)C(=CC=C2F)C(=O)O
C11H7FO2
MFCD00004011
4-fluoro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 4-fluoro, 4-fluoronaphthalenecarboxylic acid, 4-fluoro-1-naphthalenecarboxylic acid, 4-fluoro-1-naphthalene carboxylic acid, 4-fluoro-naphthalene-1-carboxylic acid, acmc-1atch, maybridge3_003028, ksc270s4j, 1-carboxy-4-fluoronaphthalene
4-fluoronaphthalene-1-carboxylic acid

Specifications

573-03-5
C11H7FO2
1 g
4-fluoro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 4-fluoro, 4-fluoronaphthalenecarboxylic acid, 4-fluoro-1-naphthalenecarboxylic acid, 4-fluoro-1-naphthalene carboxylic acid, 4-fluoro-naphthalene-1-carboxylic acid, acmc-1atch, maybridge3_003028, ksc270s4j, 1-carboxy-4-fluoronaphthalene
C1=CC=C2C(=C1)C(=CC=C2F)C(=O)O
190.173
190.18
4-Fluoro-1-naphthoic acid
223°C to 226°C
MFCD00004011
2257292
DEWIOKQDRWFLFW-UHFFFAOYSA-N
4-fluoronaphthalene-1-carboxylic acid
68448
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-346-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only