missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-naphthoic acid, 97%

Catalog No. AAA1677806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1677806 5 g
AAA1677803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1677806 Supplier Thermo Scientific Chemicals Supplier No. A1677806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 573-03-5 | C11H7FO2 | 190.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 573-03-5
Molecular Formula C11H7FO2
Molecular Weight (g/mol) 190.173
MDL Number MFCD00004011
InChI Key DEWIOKQDRWFLFW-UHFFFAOYSA-N
Synonym 4-fluoro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 4-fluoro, 4-fluoronaphthalenecarboxylic acid, 4-fluoro-1-naphthalenecarboxylic acid, 4-fluoro-1-naphthalene carboxylic acid, 4-fluoro-naphthalene-1-carboxylic acid, acmc-1atch, maybridge3_003028, ksc270s4j, 1-carboxy-4-fluoronaphthalene
PubChem CID 68448
IUPAC Name 4-fluoronaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CC=C2F)C(=O)O

Specifications

Melting Point 223°C to 226°C
Quantity 5 g
Beilstein 2257292
Formula Weight 190.18
Percent Purity 97%
Chemical Name or Material 4-Fluoro-1-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-346-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only