missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1-indanone, 97%

Catalog No. AAH5935606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5935606 5 g
AAH5935603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5935606 Supplier Thermo Scientific Chemicals Supplier No. H5935606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 699-99-0 | C9H7FO | 150.152 g/mol

It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

Solubility
Soluble in alcohol and chloroform

Notes
Store at room temperature. Keep away from oxidizing agents and heats.

Chemical Identifiers

CAS 699-99-0
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.152
MDL Number MFCD00797829
InChI Key HOMSJDBZHCPYHY-UHFFFAOYSA-N
Synonym 4-fluoro-1-indanone, 4-fluoro-2,3-dihydro-1h-inden-1-one, 4-fluoro-indan-1-one, 4-fluoroindan-1-one, 1h-inden-1-one, 4-fluoro-2,3-dihydro, 4-fluoro-2,3-dihydro-1-indenone, 4-fluoroindane-1-one, acmc-20a8cy, 4-fluoranyl-2,3-dihydroinden-1-one
PubChem CID 12174986
IUPAC Name 4-fluoro-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)F

Specifications

Melting Point 72°C to 76°C
Quantity 5 g
Solubility Information Soluble in alcohol and chloroform
Formula Weight 150.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-1-indanone
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only