accessibility menu, dialog, popup

UserName

CAS RN 699-99-0

IUPAC Name: 4-fluoro-2,3-dihydro-1H-inden-1-one
Synonyms: 4-fluoro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 150.15
Molecular Formula: C9H7FO
InChi Key: HOMSJDBZHCPYHY-UHFFFAOYSA-N
SMILES: FC1=CC=CC2=C1CCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results