missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,2-dimethoxybenzene, 98%

Catalog No. AAA1793606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1793606 5 g
AAA1793614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1793606 Supplier Thermo Scientific Chemicals Supplier No. A1793606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 398-62-9 | C8H9FO2 | 156.156 g/mol

4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,2-dimethoxybenzene is a raw material used in organic synthesis.

Solubility
Soluble in water 0.079 g/L 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 398-62-9
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00012201
InChI Key DAGKHJDZYJFWSO-UHFFFAOYSA-N
Synonym 4-fluoroveratrole, 1,2-dimethoxy-4-fluorobenzene, 3,4-dimethoxyfluorobenzene, 1-fluoro-3,4-dimethoxybenzene, 4-fluoro-1,2-dimethoxy-benzene, 3,4-dimethoxy-1-fluorobenzene, 4-fluoroveratrol, pubchem3048, acmc-1acnf, fluorobenzene, 3,4-methoxy
PubChem CID 593640
IUPAC Name 4-fluoro-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)F)OC

Specifications

Density 1.171
Boiling Point 120°C to 123°C (45 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.51
Quantity 5 g
Beilstein 2441600
Solubility Information Soluble in water 0.079g/L 25°C.
Formula Weight 156.16
Percent Purity 98%
Chemical Name or Material 4-Fluoro-1,2-dimethoxybenzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only