missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylbenzoic Acid 98.0+%, TCI America™

Catalog No. E10411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10411G Supplier TCI America Supplier No. E10411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10602-00-3
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD00168819
InChI Key SJXHLZCPDZPBPW-UHFFFAOYSA-N
PubChem CID 589706
IUPAC Name 4-ethynylbenzoic acid
SMILES C#CC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 192°C
Quantity 1 g
Formula Weight 146.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethynylbenzoic Acid