accessibility menu, dialog, popup

UserName

CAS RN 10602-00-3

IUPAC Name: 4-ethynylbenzoic acid
Synonyms: 4-ethynylbenzoic acid
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: SJXHLZCPDZPBPW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C#C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results