missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylbenzaldehyde, 95%

Catalog No. p-4228645
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432940010 1 g Glass bottle
AC432940050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432940010 Supplier Thermo Scientific Chemicals Supplier No. 432940010

CAS: 63697-96-1 | C9H6O | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63697-96-1
Molecular Formula C9H6O
Molecular Weight (g/mol) 130.15
InChI Key BGMHQBQFJYJLBP-UHFFFAOYSA-N
PubChem CID 2771645
IUPAC Name 4-ethynylbenzaldehyde
SMILES C#CC1=CC=C(C=C1)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 89.0°C to 94.5°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-ethynyl benzaldehyde, 4-ethynyl-benzaldehyde, benzaldehyde, 4-ethynyl, benzaldehyde,4-ethynyl, p-ethynylbenzaldehyde, 4-acetylenylbenzaldehyde, acmc-209x0m, 4-ethynylbenzaldehyde, benzaldehyde, 4-ethynyl-9ci
Formula Weight 130.15
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Ethynylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only