missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyltoluene 97.0+%, TCI America™

Supplier:  TCI America E0186100ML

 View more versions of this product

Catalog No. E0186100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Ethyltoluene
Colorless
C9H12
100 mL
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
CCC1=CC=C(C=C1)C
120.195
120.20
Liquid
622-96-8
161°C
MFCD00009263
3295
JRLPEMVDPFPYPJ-UHFFFAOYSA-N
1-ethyl-4-methylbenzene
12160
≥97.0% (GC)

Chemical Identifiers

622-96-8
120.195
JRLPEMVDPFPYPJ-UHFFFAOYSA-N
12160
CCC1=CC=C(C=C1)C
C9H12
MFCD00009263
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
1-ethyl-4-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0015

RTECSNumber : XT2550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions