Learn More
4-(Ethylsulfonyl)benzeneboronic acid, 98+%
CAS: 352530-24-6 | C8H11BO4S | 214.042 g/mol
Supplier: Thermo Scientific Chemicals L1781403
Description
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 4-(Ethylsulfonyl)benzeneboronic acid | |
| 135°C to 139°C | |
| MFCD02093076 | |
| 4-ethylsulfonyl phenylboronic acid, 4-ethylsulfonyl phenyl boronic acid, 4-ethanesulfonyl benzeneboronic acid, 4-ethanesulfonyl phenylboronic acid, 4-ethylsulfonylphenyl boronic acid, 4-ethylsulphonyl benzeneboronic acid, boronic acid, 4-ethylsulfonyl phenyl, 4-ethanesulfonyl phenyl boranediol, 4-ethanesulphonyl benzeneboronic acid | |
| CVBNDDGEVPNUNA-UHFFFAOYSA-N | |
| (4-ethylsulfonylphenyl)boronic acid | |
| 2773420 | |
| ≥98% |
| 352530-24-6 | |
| C8H11BO4S | |
| 1 g | |
| Slightly soluble in water. | |
| B(C1=CC=C(C=C1)S(=O)(=O)CC)(O)O | |
| 214.042 | |
| 214.05 |
Chemical Identifiers
| 352530-24-6 | |
| 214.042 | |
| CVBNDDGEVPNUNA-UHFFFAOYSA-N | |
| 2773420 | |
| B(C1=CC=C(C=C1)S(=O)(=O)CC)(O)O |
| C8H11BO4S | |
| MFCD02093076 | |
| 4-ethylsulfonyl phenylboronic acid, 4-ethylsulfonyl phenyl boronic acid, 4-ethanesulfonyl benzeneboronic acid, 4-ethanesulfonyl phenylboronic acid, 4-ethylsulfonylphenyl boronic acid, 4-ethylsulphonyl benzeneboronic acid, boronic acid, 4-ethylsulfonyl phenyl, 4-ethanesulfonyl phenyl boranediol, 4-ethanesulphonyl benzeneboronic acid | |
| (4-ethylsulfonylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only