missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylresorcinol 98.0+%, TCI America™

Catalog No. p-7126299
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D05795G 5 g
D057925G 25 g
2 options

Catalog No. D05795G

Supplier: TCI America D05795G

Only null left

Chemical Identifiers

CAS 2896-60-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002283
InChI Key VGMJYYDKPUPTID-UHFFFAOYSA-N
Synonym 4-ethylresorcinol, 1,3-benzenediol, 4-ethyl, 6-ethylresorcinol, resorcinol, 4-ethyl, 1,3-dihydroxy-4-ethylbenzene, 4-ethyl resorcinol, 2,4-dihydroxy-1-ethylbenzene, unii-8ye81t06d5, 4-ethyl-benzene-1,3-diol, 4-ethyl-resorcinol
PubChem CID 17927
IUPAC Name 4-ethylbenzene-1,3-diol
SMILES CCC1=C(C=C(C=C1)O)O

Specifications

Melting Point 98°C
Boiling Point 155°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethylresorcinol
EINECSNumber (4)-0057
TSCA Yes