missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylpyridine, 98%

Catalog No. AAA1288318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1288318 50 g
AAA1288330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1288318 Supplier Thermo Scientific Chemicals Supplier No. A1288318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 536-75-4 | C7H9N | 107.156 g/mol

4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

Solubility
Slightly soluble in water (34 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 536-75-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006450
InChI Key VJXRKZJMGVSXPX-UHFFFAOYSA-N
Synonym pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine
PubChem CID 10822
IUPAC Name 4-ethylpyridine
SMILES CCC1=CC=NC=C1

Specifications

Melting Point -91°C
Density 0.942
pH 10
Boiling Point 167°C to 168°C
Flash Point 52°C (126°F)
Odor Unpleasant
Refractive Index 1.501
Quantity 50 g
UN Number UN1993
Beilstein 106481
Merck Index 14,3849
Solubility Information Slightly soluble in water (34g/L at 20°C).
Formula Weight 107.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Ethylpyridine
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement

H226-H302-H312-H332-H315-H319-H335

Flammable liquid and vapour.

Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-646-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only