missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals

$170.88 - $170.88

Chemical Identifiers

CAS 18856-63-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00041101
InChI Key XXLWCGRHJFEMQH-UHFFFAOYSA-N
Synonym 4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
PubChem CID 140438
IUPAC Name 1-ethyl-4-isothiocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2435309
View Documents
Thermo Scientific Chemicals
B2435309
10 g
Each for $170.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18856-63-8
163.24
XXLWCGRHJFEMQH-UHFFFAOYSA-N
140438
CCC1=CC=C(C=C1)N=C=S
C9H9NS
MFCD00041101
4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
1-ethyl-4-isothiocyanatobenzene

Specifications

18856-63-8
245°C
Pungent
1.6205
10 g
2802384
4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
CCC1=CC=C(C=C1)N=C=S
163.24
163.24
4-Ethylphenyl isothiocyanate
1.075
>110°C (230°F)
C9H9NS
MFCD00041101
UN2922
Moisture sensitive
XXLWCGRHJFEMQH-UHFFFAOYSA-N
1-ethyl-4-isothiocyanatobenzene
140438
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only