missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylnitrobenzene 99.0+%, TCI America™

Catalog No. p-7128866
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E015425G 25 g
E0154500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E015425G Supplier TCI America Supplier No. E015425G

Chemical Identifiers

CAS 100-12-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007385
InChI Key RESTWAHJFMZUIZ-UHFFFAOYSA-N
Synonym 4-ethylnitrobenzene, benzene, 1-ethyl-4-nitro, p-ethylnitrobenzene, p-nitroethylbenzene, p-nitrophenylethane, benzene,1-ethyl-4-nitro, 4-ethyl-1-nitrobenzene, 1-ethyl-4-nitro-benzene, ccris 3100, 4-nitroethylbenzene
PubChem CID 7480
IUPAC Name 1-ethyl-4-nitrobenzene
SMILES CCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point -18°C
Boiling Point 242°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylnitrobenzene
EINECSNumber (3)-4058
TSCA Yes