missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylcyclohexanol (cis- and trans- mixture) 97.0+%, TCI America™
Supplier: TCI America E036625ML
Spécifications
| 4-Ethylcyclohexanol (cis- and trans- mixture) | |
| Yellow | |
| MFCD00001450,MFCD00070694 | |
| 4-ethylcyclohexanol, trans-4-ethylcyclohexanol, cyclohexanol, 4-ethyl, cis-4-ethylcyclohexanol, cyclohexanol, 4-ethyl-, trans, 4-ethylcyclohexanol,c&t, 4-ethylcyclohexanol mixture of cis and trans, trans-4-ethyl cyclohexanol, ethyl cyclohexanol, cyclohexanol, 4-ethyl-, mixture of cis and trans | |
| CCC1CCC(O)CC1 | |
| 128.22 | |
| 128.22 | |
| Liquid |
| 4534-74-1 | |
| C8H16O | |
| 25 mL | |
| RVTKUJWGFBADIN-UHFFFAOYSA-N | |
| 4-ethylcyclohexan-1-ol | |
| 78293 | |
| ≥97.0% (GC) |
Identifiants chimiques
| 4534-74-1 | |
| 128.22 | |
| RVTKUJWGFBADIN-UHFFFAOYSA-N | |
| 78293 | |
| CCC1CCC(O)CC1 |
| C8H16O | |
| MFCD00001450,MFCD00070694 | |
| 4-ethylcyclohexanol, trans-4-ethylcyclohexanol, cyclohexanol, 4-ethyl, cis-4-ethylcyclohexanol, cyclohexanol, 4-ethyl-, trans, 4-ethylcyclohexanol,c&t, 4-ethylcyclohexanol mixture of cis and trans, trans-4-ethyl cyclohexanol, ethyl cyclohexanol, cyclohexanol, 4-ethyl-, mixture of cis and trans | |
| 4-ethylcyclohexan-1-ol |
Sécurité et manipulation
missing translation for 'einecsNumber' : (3)-2301
missing translation for 'rtecsNumber' : GV9050000
missing translation for 'tsca' : No