CAS RN 4534-74-1
CAS RN 4534-74-1
IUPAC Name:
4-ethylcyclohexan-1-ol
Synonyms:
4-ethylcyclohexan-1-ol
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
RVTKUJWGFBADIN-UHFFFAOYSA-N
SMILES:
CCC1CCC(O)CC1
1
–
1
of
1
results
1
4-Ethylcyclohexanol (cis- and trans- mixture) 97.0+%, TCI America™
CAS: 4534-74-1 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.22 MDL Number: MFCD00001450,MFCD00070694 InChI Key: RVTKUJWGFBADIN-UHFFFAOYSA-N Synonym: 4-ethylcyclohexanol,trans-4-ethylcyclohexanol,cyclohexanol, 4-ethyl,cis-4-ethylcyclohexanol,cyclohexanol, 4-ethyl-, trans,4-ethylcyclohexanol,c&t,4-ethylcyclohexanol mixture of cis and trans,trans-4-ethyl cyclohexanol,ethyl cyclohexanol,cyclohexanol, 4-ethyl-, mixture of cis and trans PubChem CID: 78293 IUPAC Name: 4-ethylcyclohexan-1-ol SMILES: CCC1CCC(O)CC1