missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcatechol, 98%

Catalog No. p-7081877
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12048MD 250 mg
AAA1204803 1 g
AAA1204806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12048MD Supplier Thermo Scientific Chemicals Supplier No. A12048MD
Only null left

CAS: 1124-39-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1124-39-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00015847
InChI Key HFLGBNBLMBSXEM-UHFFFAOYSA-N
Synonym 4-ethylcatechol, 4-ethylpyrocatechol, 4-ethyl-1,2-benzenediol, unii-574jv8byr2, 1,2-benzenediol, 4-ethyl, benzenediol, 4-ethyl, 3,4-dihydroxyethylbenzene, ethylcatechol, 4-ethyl catechol, 4-ethylc atechol
PubChem CID 70761
IUPAC Name 4-ethylbenzene-1,2-diol
SMILES CCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 37°C to 40°C
Boiling Point 105°C to 107°C (0.5 mmHg)
Quantity 250 mg
Beilstein 2042062
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Ethylcatechol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-397-5
TSCA No

RUO – Research Use Only