missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. E07045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07045G 5 g
E070425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07045G Supplier TCI America Supplier No. E07045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 768-59-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004666
InChI Key YSLBFFIVJGJBSA-UHFFFAOYSA-N
PubChem CID 13034
IUPAC Name (4-ethylphenyl)methanol
SMILES CCC1=CC=C(C=C1)CO

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 g
Synonym 4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol #
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzyl Alcohol
EINECSNumber (3)-1049
TSCA No