missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylbenzyl alcohol, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.028 |
| Boiling Point | 115°C to 117°C (9 mmHg) |
| Refractive Index | 1.5275 |
| Quantity | 1 g |
| Synonym | 4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol # |
| Formula Weight | 136.19 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-Ethylbenzyl alcohol |
Safety and Handling
| EINECSNumber | 292-198-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only