missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl alcohol, 99%

Catalog No. AAH3045906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3045906 5 g
AAH3045903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3045906 Supplier Thermo Scientific Chemicals Supplier No. H3045906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 768-59-2 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-59-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004666
InChI Key YSLBFFIVJGJBSA-UHFFFAOYSA-N
PubChem CID 13034
IUPAC Name (4-ethylphenyl)methanol
SMILES CCC1=CC=C(C=C1)CO

Specifications

Density 1.028
Boiling Point 115°C to 117°C (9 mmHg)
Refractive Index 1.5275
Quantity 5 g
Synonym 4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol #
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 4-Ethylbenzyl alcohol
EINECSNumber 292-198-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only