missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzenesulfonyl Chloride 95.0+%, TCI America™

Catalog No. E006525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E006525G Supplier TCI America Supplier No. E006525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16712-69-9
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00041480
InChI Key LACFLXDRFOQEFZ-UHFFFAOYSA-N
Synonym 4-ethylbenzene-1-sulfonyl chloride, p-ethylbenzenesulfonyl chloride, 4-ethylbenzenesulphonyl chloride, 4-ethyl-benzenesulfonyl chloride, ethylbenzenesulfonylchloride, 4-ethylbenzenesulfonylchloride, chloro 4-ethylphenyl sulfone, pubchem5133, acmc-209dvn, 4-ethylphenylsulfonyl chloride
PubChem CID 570959
IUPAC Name 4-ethylbenzenesulfonyl chloride
SMILES CCC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 204.67
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzenesulfonyl Chloride