accessibility menu, dialog, popup

UserName

H3CSOOCl

CAS RN 16712-69-9

IUPAC Name: 4-ethylbenzene-1-sulfonyl chloride
Synonyms: 4-ethylbenzenesulfonyl chloride
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: LACFLXDRFOQEFZ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)